Learn More
Aviva Systems Biology Corp AGPAT2 Antibody - C-terminal region (ARP44636_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGPAT2. It was validated on Western Blot. . Alternate names: 1-AGPAT2, BSCL, BSCL1, LPAAB, LPAAT-betaApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Yeast
Catalog No. 50-285-155