Learn More
Aviva Systems Biology Corp AGPAT3 Antibody - middle region (ARP60800_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60800_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGPAT3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human AGPAT3. Alternate names: LPAAT-GAMMA1, MGC4604ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-287-168