Learn More
Aviva Systems Biology Corp AGPAT4 Antibody - middle region (ARP60821_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60821_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGPAT4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human AGPAT4. Alternate names: 1-AGPAT4, LPAAT-delta, dJ473J16.2, RP3-473J16.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-287-181