Learn More
Aviva Systems Biology Corp GPAT4 Antibody - middle region (ARP60802_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60802_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGPAT6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human AGPAT6. Alternate names: DKFZp586M1819, LPAAT-zeta, LPAATZ, TSARG7, 1-AGPAT 6ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-169