Learn More
Aviva Systems Biology Corp GPAT3 Antibody - middle region (ARP50103_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50103_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Agpat9. It was validated on Western Blot. . Alternate names: MGC114395ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-488