Learn More
Aviva Systems Biology Corp AGR2 Antibody - middle region (ARP42290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AGR2. Alternate names: AG2, GOB-4, HAG-2, XAG-2, PDIA17ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58174