Learn More
ROMER LABS INC AgraQuant Gluten G12 ELISA Test Kit, 96 wells, quantitative, detection limit 2 ppm, quantitation range 4-200 ppm, ready-to-use reagents, includes extraction buffer
Supplier:  ROMER LABS INC 10001994
Encompass
AgraQuant Gluten G12 ELISA Test Kit, 96 wells, quantitative, detection limit 2 ppm, quantitation range 4-200 ppm, ready-to-use reagents, includes extraction buffer
Catalog No. NC1984116