Learn More
Roboz Surgical AGRICOLA RETRACTOR 3X3 BLNT EA
SDP
Catalog No. NC0301827
Encompass
Agricola Retractor; 3 X 3 Blunt Prongs; 4mm Depth; 25mm Spread; Length 1-1/2 in.
Catalog No. NC0301827 Supplier Roboz Surgical Supplier No. RS6501
Agricola Retractor; 3 X 3 Blunt Prongs; 4mm Depth; 25mm Spread; Length 1-1/2 in.