Learn More
World Precision Instrument Agricola Self-Retaining Retractor, 4cm, 3x3 Sharp Prongs
SDP
Catalog No. 502521396
Encompass
  • 4cm (1.6") Long
  • 3 x 3 Sharp Prongs
  • Small, self-retaining retractor, perfect for small animal surgery and dissection
  • Maximum spread: 2.5cm
  • Stainless steel
Catalog No. 50-252-1396 Supplier World Precision Instrument Supplier No. 501846
  • 4cm (1.6") Long
  • 3 x 3 Sharp Prongs
  • Small, self-retaining retractor, perfect for small animal surgery and dissection
  • Maximum spread: 2.5cm
  • Stainless steel