Learn More
Enzo Life Sciences Agrin monoclonal antibody (Agr-131) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIAGR530D
Encompass_Preferred
Clone: Agr-131. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant rat agrin (C-terminal fragment). UniProt: P25304. Source: Purified from mouse ascites. Species Reactivity: Mouse, Rat. Applications: ICC, IHC (PS), WB, Microarray. Recommended dilutions: Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-658-963