Learn More
Medchemexpress LLC Agrin Human CHO H 100ug | 100ug
SDP
Catalog No. 5000194730
Encompass_Preferred
Agrin is critical for neuromuscular junction development and was discovered in mouse knockout studies It contains laminin G Kazal-type serpin and epidermal growth factor domains Agrin Protein Human (CHO His) is the recombinant human-derived Agrin protein expressed by CHO with N-6 His labeled tag
Catalog No. 50-001-94730 Supplier Medchemexpress LLC Supplier No. HYP79236100UG
Agrin is critical for neuromuscular junction development and was discovered in mouse knockout studies It contains laminin G Kazal-type serpin and epidermal growth factor domains Agrin Protein Human (CHO His) is the recombinant human-derived Agrin protein expressed by CHO with N-6 His labeled tag