Learn More
Medchemexpress LLC Agrin Human CHO H 500ug | 500ug
SDP
Supplier:  Medchemexpress LLC HYP79236500UG
Encompass_Preferred
Agrin is critical for neuromuscular junction development and was discovered in mouse knockout studies It contains laminin G Kazal-type serpin and epidermal growth factor domains Agrin Protein Human (CHO His) is the recombinant human-derived Agrin protein expressed by CHO with N-6 His labeled tag
Catalog No. 50-001-94675