Learn More
Enzo Life Sciences Agrin monoclonal antibody (Agr-131) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIAGR530F
Encompass_Preferred
Clone: Agr-131. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant rat agrin (C-terminal fragment). Source: Purified from mouse ascites. Species reactivity: Mouse, Rat. Applications: ICC, IHC (PS), WB Microarray. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7173