Learn More
Enzo Life Sciences Agrin monoclonal antibody (Agr-247) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIAGR540D
Encompass_Preferred
Clone: Agr-247. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat agrin (C-terminal fragment). UniProt ID: P25304. Source: Purified from mouse ascites. Species reactivity: Mouse, Rat. Applications: IHC, WB. Recommended Dilutions: Immunohistochemistry (10µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7174