Learn More
Enzo Life Sciences Agrin monoclonal antibody (Agr-33) (1mg)
SDP
Supplier:  Enzo Life Sciences ADIAGR510J
Encompass_Preferred
Clone: Agr-33. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat agrin (C-terminal fragment). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Mouse, Rat. Applications: ICC, IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7172