Learn More
Cayman Chemical Agrmona plosa Agrmol B 10mg
SDP
Supplier:  Cayman Chemical 3499610 MG
Encompass_Preferred
A polyphenol with adipogenesis inhibitory and anticancer activities; induces nuclear translocation of SIRT1 and reduces expression of PPARγ, C/EBPα, and FASN in 3T3-L1 adipocytes at 10 µM; reduces lipid droplet accumulation in 3T3-L1 adipocytes (IC50 = 3.35 µM); cytotoxic to PC3 prostate and A549 lung cancer cells in vitro; reduces tumor volume in a PC3 mouse xenograft model at 10 mg/kg
Catalog No. 50-291-1017