Learn More
ATCC AGS; Gastric Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382867
Encompass
AGS is a cell line exhibiting epithelial morphology that was isolated in 1979 from the stomach tissue of a 54-year-old, White, female patient with gastric adenocarcinoma.
Catalog No. 50-238-2867 Supplier ATCC Supplier No. CRL1739
AGS is a cell line exhibiting epithelial morphology that was isolated in 1979 from the stomach tissue of a 54-year-old, White, female patient with gastric adenocarcinoma.