Learn More
Aviva Systems Biology Corp AGTPBP1 Antibody - N-terminal region (ARP55205_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55205_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGTPBP1. It was validated on Western Blot. AGTPBP1,ATP/GTP binding protein 1,NNA1; KIAA1035; DKFZp686M20191; AGTPBP1,cytosolic carboxypeptidase 1,cytosolic carboxypeptidase 1,OTTHUMP00000021580,ATP/GTP-binding protein 1,nervous system nuclear protein induced by axotomyThe immunogen is a synthetic peptide directed towards the N terminal region of human AGTPBP1. Alternate names: DKFZp686M20191, KIAA1035, NNA1, CCP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-014