Learn More
Aviva Systems Biology Corp AGTR1 Antibody - N-terminal region (ARP42198_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42198_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGTR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AGTR1. Alternate names: AG2S, AGTR1A, AGTR1B, AT1, AT1B, AT2R1, AT2R1A, AT2R1B, HAT1R, AT1RApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53327