Learn More
Eurofins Discoverx ChemiScreen AT2 Angiotensin II Receptor Membrane Preparation, 1 vial. (HTS208M)
Catalog No. 502006056
Encompass
The ChemiScreen AT2 Angiotensin II Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of AGTR2 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-6056 Supplier Eurofins Discoverx Supplier No. HTS208M
The ChemiScreen AT2 Angiotensin II Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of AGTR2 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.