Learn More
Aviva Systems Biology Corp AGXT2 Antibody - N-terminal region (ARP48953_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48953_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGXT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AGXT2. Alternate names: AGT2, DAIBATApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59973