Learn More
Aviva Systems Biology Corp AGXT2L2 Antibody - C-terminal region (ARP47730_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47730_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGXT2L2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AGXT2L2. Alternate names: MGC117348, MGC15875, MGC45484, AGXT2L2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53528