Learn More
Enzo Life Sciences AH-6809 (25 mg). CAS: 33458-93-4
SDP
Catalog No. 502011715
Encompass_Preferred
Selective prostaglandin E2 receptor (EP1) antagonist. Useful at concentrations up to 10 µM, at which point it starts to block PGD2 and TXA2 receptors. Purity: ≥98% (TLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-1715 Supplier Enzo Life Sciences Supplier No. BMLRA1100025
Selective prostaglandin E2 receptor (EP1) antagonist. Useful at concentrations up to 10 µM, at which point it starts to block PGD2 and TXA2 receptors. Purity: ≥98% (TLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.