Learn More
TARGETMOL CHEMICALS INC AH-7614 25MG
SDP
Catalog No. 502580985
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AH-7614 (AH 7614) is a selective and potent free fatty acid receptor 4 (FFA4/GPR120) antagonist. purity: 99%
Catalog No. 50-258-0985 Supplier TARGETMOL CHEMICALS INC Supplier No. T2202725MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AH-7614 (AH 7614) is a selective and potent free fatty acid receptor 4 (FFA4/GPR120) antagonist. purity: 99%