Learn More
Medchemexpress LLC AH-D peptide TFA | 98.8% | 3283.75 | 5 MG
SDP
Catalog No. 5000424067
Encompass_Preferred
AH-D peptide TFA is an antiviral peptide that selectively disrupts membrane structures within the size range of exosomes, inducing T-EXO depletion and enhancing cancer immunotherapy.
  • Antiviral peptide
  • Disrupts membrane structures within exosome size range
  • Induces T-EXO depletion
  • Enhances cancer immunotherapy
  • Solid, white to off-white appearance
  • Store sealed, away from moisture
  • Powder storage at -80°C for 2 years, -20°C for 1 year
  • In solvent storage at -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-24067 Supplier Medchemexpress LLC Supplier No. HYP10862A5MG
AH-D peptide TFA is an antiviral peptide that selectively disrupts membrane structures within the size range of exosomes, inducing T-EXO depletion and enhancing cancer immunotherapy.
  • Antiviral peptide
  • Disrupts membrane structures within exosome size range
  • Induces T-EXO depletion
  • Enhances cancer immunotherapy
  • Solid, white to off-white appearance
  • Store sealed, away from moisture
  • Powder storage at -80°C for 2 years, -20°C for 1 year
  • In solvent storage at -80°C for 6 months, -20°C for 1 month