Learn More
AnaSpec AH1 SEQUENCE (6-14) 1MG
Supplier:  AnaSpec AS64798
Encompass
AH1 Sequence (6-14)[SPSYVYHQF] ; MW 1127.2; (1 mg) This AH1 sequence (amino acids 6 to 14) is the H2-Ld-restricted epitope derived from gp-70 (endogenous retroviral gene product envelope glycoprotein 70), which is expressed in CT26 (colon carcinoma cells) and numerous other tumor cell lines. AH
Catalog No. NC0950043