Learn More
Aviva Systems Biology Corp Ahcy Antibody - N-terminal region (ARP51769_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51769_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ahcy. It was validated on Western Blot. . Alternate names: AA987153, AL024110, CuBP, MGC102079, MGC118063, MGC19228, SAHHApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-590