Learn More
Aviva Systems Biology Corp AHCY Antibody - N-terminal region (ARP51768_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51768_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AHCY. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AHCY. Alternate names: SAHHApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60610