Learn More
Aviva Systems Biology Corp AHCYL1 Antibody - N-terminal region (ARP33131_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33131_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AHCYL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AHCYL1. Alternate names: DCAL, IRBIT, PRO0233, XPVKONAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54892