Learn More
New England Biolabs, Inc. AhdI – 1000 units
SDP
Catalog No. 50812290
Encompass
A restriction endonuclease that recognizes the sequence GACNN_N^NNGTC.
Catalog No. 50-812-290 Supplier New England Biolabs, Inc. Supplier No. R0584S
A restriction endonuclease that recognizes the sequence GACNN_N^NNGTC.