Learn More
Aviva Systems Biology Corp Ahi1 Antibody - C-terminal region (ARP56992_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56992_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ahi1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Ahi-1, Ahi1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-252