Learn More
TARGETMOL CHEMICALS INC AHN 1-055 HYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3942925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AHN 1-055 hydrochloride (3(alpha)-Bis-(4-fluorophenyl) Methoxytropane hydrochloride) is an inhibitor of dopamine transporter (DAT) and dopamine uptake(IC50 = 71 nM). purity: 99%
Catalog No. 50-258-0053