Learn More
Medchemexpress LLC HY-101315 5mg Medchemexpress, AHN 1-055 (hydrochloride) CAS:202646-03-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1013155MG
Encompass_Preferred
Medchemexpress, HY-101315 5mg AHN 1-055 (hydrochloride) CAS:202646-03-5 AHN 1-055 hydrochloride is a dopamine uptake inhibitor, with an IC50 of 71 nM. AHN 1-055 hydrochloride binds with high affinity to the dopamine transporter (DAT) and may serve as leads for the development of agentia to treat cocaine abuse. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0627