Learn More
Sino Biological Anti-AHNAK2/AHNAK nucleoprotein 2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 203703T10100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human AHNAK2/AHNAK nucleoprotein 2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-228-2940