Learn More
Aviva Systems Biology Corp AHR Antibody - N-terminal region (ARP32243_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32243_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AHR. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human AHR. Alternate names: bHLHe76ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54495