Learn More
Aviva Systems Biology Corp AHR Antibody - N-terminal region (ARP31635_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31635_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AHR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AHR. Alternate names: bHLHe76ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54289