Learn More
Enzo Life Sciences Aryl hydrocarbon receptor monoclonal antibody (RPT1) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804421R100
Encompass_Preferred
Clone: RPT1. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 12-17 (R12KRRKP17) and aa 22-31 (V22KPIPAEGIK31) of mouse AhR (aryl hydrocarbon receptor). Species reactivity: Human, Mouse, Rat. Applications: IF, IHC (PS), IP, WB. Formulation: Liquid. Ascites containing 0.05% sodium azide.
Catalog No. 50-200-9905