Learn More
Enzo Life Sciences Aryl hydrocarbon receptor monoclonal antibody (RPT9) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804423R100
Encompass_Preferred
Clone: RPT9. Isotype: IgG3. Immunogen: Synthetic peptide corresponding to aa 12-17 (R12KRRKP17) and aa 22-31 (V22KPIPAEGIK31) of mouse AhR (aryl hydrocarbon receptor). Species reactivity: Human, Mouse, Rat. Applications: Flow Cytometry, ICC, IF, IHC (FS), IP, WB Gel Shift, ChIP. Purity: Protein A-affinity purified. Formulation: Liquid. Contains 0.05% sodium azide.
Catalog No. 50-200-9907