Learn More
Enzo Life Sciences AhR polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA5500100
Encompass_Preferred
Immunogen: Recombinant B-1 AhR (aa 1-402). Source: Whole rabbit serum. Species reactivity: Mouse, Rat. Applications: IP, WB. Formulation: Liquid. Neat serum.
Catalog No. 50-201-1848