Learn More
ABclonal Technology AHR Rabbit pAb
SDP
Catalog No. 5000107091
Encompass_Preferred
The protein encoded by this gene is a ligand-activated helix-loop-helix transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. This receptor has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450. Before ligand binding, the encoded protein is sequestered in the cytoplasm, upon ligand binding, this protein moves to the nucleus and stimulates transcription of target genes.
Catalog No. 50-001-07091 Supplier ABclonal Technology Supplier No. A14511000UL
The protein encoded by this gene is a ligand-activated helix-loop-helix transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. This receptor has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450. Before ligand binding, the encoded protein is sequestered in the cytoplasm, upon ligand binding, this protein moves to the nucleus and stimulates transcription of target genes.