Learn More
Aviva Systems Biology Corp AHRR Antibody - middle region (ARP39438_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AHRR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AHRR. Alternate names: AHH, AHHR, KIAA1234, MGC167813, MGC176630, bHLHe77ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53139