Learn More
Aviva Systems Biology Corp AHSG Antibody - N-terminal region (ARP33815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AHSG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AHSG. Alternate names: A2HS, AHS, FETUA, HSGAApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-55159