Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-2-HS-glycoprotein (fetuin-A), recombinant mouse, His tag | >95.0% | 100UG
SDP

Catalog No. 5000175551
Encompass_Preferred

AHSG/Fetuin-A Protein, Mouse (HEK293, His) is a recombinant phosphorylated glycoprotein produced in HEK293 cells and supplied as a lyophilized His-tagged protein for research use. The protein exhibits mammalian glycosylation, high purity, and low endotoxin, and includes recommended reconstitution and storage conditions to preserve activity and reproducibility.

  • Expressed in HEK293 cells to retain mammalian post-translational modifications.
  • C-terminal His tag for detection and purification applications.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level less than 1 EU/μg for low immunogenic interference.
  • Lyophilized formulation with defined buffer for stability and handling.
  • Recommended storage at -20°C and aliquoting for extended stability.
  • Not recommended to reconstitute below 100 μg/mL; add carrier protein for long-term storage.

Catalog No. 50-001-75551 Supplier Medchemexpress LLC Supplier No. HYP70339100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AHSG/Fetuin-A Protein, Mouse (HEK293, His) is a recombinant phosphorylated glycoprotein produced in HEK293 cells and supplied as a lyophilized His-tagged protein for research use. The protein exhibits mammalian glycosylation, high purity, and low endotoxin, and includes recommended reconstitution and storage conditions to preserve activity and reproducibility.

  • Expressed in HEK293 cells to retain mammalian post-translational modifications.
  • C-terminal His tag for detection and purification applications.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level less than 1 EU/μg for low immunogenic interference.
  • Lyophilized formulation with defined buffer for stability and handling.
  • Recommended storage at -20°C and aliquoting for extended stability.
  • Not recommended to reconstitute below 100 μg/mL; add carrier protein for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.