Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-2-HS-glycoprotein (Fetuin-A), mouse, recombinant (HEK293, His tag) | 9014-81-7 | >95.0% | 5UG
SDP

Catalog No. 5000176058
Encompass_Preferred

Recombinant mouse AHSG (Fetuin-A) protein expressed in HEK293 cells with a C-terminal His tag. AHSG is a phosphorylated serum glycoprotein primarily produced by hepatocytes; this recombinant preparation is supplied for research applications in biochemical and functional studies.

  • Expressed in HEK293 cells with C-terminal His tag.
  • Phosphorylated glycoprotein derived from mouse AHSG.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight 45-71 kDa.
  • Available in multiple microgram packaging options.
  • Suitable for biochemical assays, antibody validation, and functional studies.

Catalog No. 50-001-76058 Supplier Medchemexpress LLC Supplier No. HYP703395UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse AHSG (Fetuin-A) protein expressed in HEK293 cells with a C-terminal His tag. AHSG is a phosphorylated serum glycoprotein primarily produced by hepatocytes; this recombinant preparation is supplied for research applications in biochemical and functional studies.

  • Expressed in HEK293 cells with C-terminal His tag.
  • Phosphorylated glycoprotein derived from mouse AHSG.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight 45-71 kDa.
  • Available in multiple microgram packaging options.
  • Suitable for biochemical assays, antibody validation, and functional studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.