Learn More
Cayman Chemical AHU377calcIum salt 100mg
SDP
Catalog No. 502985821
Encompass_Preferred
A prodrug form of LBQ657; increases plasma levels of ANF in concious rats and anesthetized dogs at 10 and 30 mg/kg, respectively; increases ANF-induced urinary sodium excretion in anesthetized rats at 30 mg/kg
Catalog No. 50-298-5821 Supplier Cayman Chemical Supplier No. 21473100 MG
A prodrug form of LBQ657; increases plasma levels of ANF in concious rats and anesthetized dogs at 10 and 30 mg/kg, respectively; increases ANF-induced urinary sodium excretion in anesthetized rats at 30 mg/kg