Learn More
Cayman Chemical AHU377 (calcium salt), Purity: ≥98%, CAS: 1369773-39-6, 50 mg
SDP
Catalog No. 502987384
Encompass_Preferred
A prodrug form of LBQ657; increases plasma levels of ANF in concious rats and anesthetized dogs at 10 and 30 mg/kg, respectively; increases ANF-induced urinary sodium excretion in anesthetized rats at 30 mg/kg
Catalog No. 50-298-7384 Supplier Cayman Chemical Supplier No. 2147350 MG
A prodrug form of LBQ657; increases plasma levels of ANF in concious rats and anesthetized dogs at 10 and 30 mg/kg, respectively; increases ANF-induced urinary sodium excretion in anesthetized rats at 30 mg/kg