Learn More
Cayman Chemical α-Hydroxyglutaric Acid (sodium salt), Purity: ≥95%, CAS: 40951-21-1, 100 mg
SDP
Catalog No. 502969901
Encompass_Preferred
An α-hydroxy acid; levels are increased in 2-hydroxyglutaric aciduria; can be metabolized from isocitrate by mutated forms of IDH1 and IDH2; competitively inhibits α-ketoglutarate-dependent dioxygenases, including histone lysine demethylases and DNA hydroxylases
Catalog No. 50-296-9901 Supplier Cayman Chemical Supplier No. 16374100 MG
An α-hydroxy acid; levels are increased in 2-hydroxyglutaric aciduria; can be metabolized from isocitrate by mutated forms of IDH1 and IDH2; competitively inhibits α-ketoglutarate-dependent dioxygenases, including histone lysine demethylases and DNA hydroxylases