Learn More
Cayman Chemical aHydroxyglutarIc AcIdodIu 10mg
SDP
Catalog No. 502972322
Encompass_Preferred
An α-hydroxy acid; levels are increased in 2-hydroxyglutaric aciduria; can be metabolized from isocitrate by mutated forms of IDH1 and IDH2; competitively inhibits α-ketoglutarate-dependent dioxygenases, including histone lysine demethylases and DNA hydroxylases
Catalog No. 50-297-2322 Supplier Cayman Chemical Supplier No. 1637410 MG
An α-hydroxy acid; levels are increased in 2-hydroxyglutaric aciduria; can be metabolized from isocitrate by mutated forms of IDH1 and IDH2; competitively inhibits α-ketoglutarate-dependent dioxygenases, including histone lysine demethylases and DNA hydroxylases