Learn More
Cayman Chemical ahydroy Farnsyl phsphonIc 10mg
SDP
Catalog No. 502895538
Encompass_Preferred
α-hydroxy Farnesyl phosphonic acid is a nonhydrolyzable analog of farnesyl pyrophosphate which acts as a competitive inhibitor of farnesyl transferase (FTase).{1199,5456} At concentrations greater than 1 µM, α-hydroxy farnesyl phosphonic acid inhibits the processing of Ras in Ha-ras-transformed NIH3T3 cells.{1196}
Catalog No. 50-289-5538 Supplier Cayman Chemical Supplier No. 6342010 MG
α-hydroxy Farnesyl phosphonic acid is a nonhydrolyzable analog of farnesyl pyrophosphate which acts as a competitive inhibitor of farnesyl transferase (FTase).{1199,5456} At concentrations greater than 1 µM, α-hydroxy farnesyl phosphonic acid inhibits the processing of Ras in Ha-ras-transformed NIH3T3 cells.{1196}