Learn More
Across International Ai 200°C 0.5 Cu Ft 4 Shelves Max Forced Air Convection Oven 110V
SDP
Catalog No. 502251213
Encompass_Preferred
Ai 0.5 cu ft forced air ovens use a microcontroller to enhance precision, accuracy and uniformity. Memory function retains all your settings when power is down and resumes previous oven operation when power is back up. Alarm can be set to trigger when current temperature is outside of desired range.
This item is not returnable. View return policy
Catalog No. 50-225-1213 Supplier Across International Supplier No. FO19013
Ai 0.5 cu ft forced air ovens use a microcontroller to enhance precision, accuracy and uniformity. Memory function retains all your settings when power is down and resumes previous oven operation when power is back up. Alarm can be set to trigger when current temperature is outside of desired range.